4-Chloro-6-(trifluoromethyl)-1H-pyrazolo[3,4-d]pyrimidine ≥95%

Supplier: Matrix Scientific

072503-250MG 072503-1G
102525-184EA 1110.77 USD
102525-184 102525-186
4-Chloro-6-(trifluoromethyl)-1H-pyrazolo[3,4-d]pyrimidine ≥95%
4-Chloro-6-(trifluoromethyl)-1H-pyrazolo[3,4-d]pyrimidine

Hazard: Irritant

Formula: C₆H₂ClF₃N₄
CAS Number: 1780-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR