2-Amino-8-methoxyquinazoline ≥95%

Supplier: Matrix Scientific

072501-250MG 072501-1G
102525-176EA 924.62 USD
102525-176 102525-178
2-Amino-8-methoxyquinazoline ≥95%
2-Amino-8-methoxyquinazoline

Hazard: Irritant

Formula: C₉H₉N₃O
MW: 175.1900 g/mol
MDL Number: MFCD09999246
CAS Number: 708-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR