Ethyl-1-benzyl-4-cyanopiperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-benzyl-4-cyanopiperidine-4-carboxylate

072498-250MG 072498-1G
102525-162EA 640 USD
102525-162 102525-164
Ethyl-1-benzyl-4-cyanopiperidine-4-carboxylate ≥95%
Ethyl-1-benzyl-4-cyanopiperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₂
MW: 272.3400 g/mol
MDL Number: MFCD09999241
CAS Number: 123730-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR