8-Bromo-2H-chromen-2-one ≥95%

Supplier: Matrix Scientific

072497-250MG 072497-1G 072497-5G
102525-156EA 640 USD
102525-156 102525-158 102525-160
8-Bromo-2H-chromen-2-one ≥95%
8-Bromo-2H-chromen-2-one

Hazard: Irritant

Formula: C₉H₅BrO₂
MDL Number: MFCD09999238
CAS Number: 33491-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR