3-Bromo-2-hydroxybenzoic acid ≥95%

Supplier: Matrix Scientific

072496-1G 072496-5G
102525-152EA 38.46 USD
102525-152 102525-154
3-Bromo-2-hydroxybenzoic acid ≥95%
3-Bromo-2-hydroxybenzoic acid

Hazard: Irritant

Formula: C7H5BrO3
MDL Number: MFCD03265705
CAS Number: 3883-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR