5-Bromo-3,7-dihydro-4H-pyrrolo[2,3-d]pyrimidin-4-one ≥95%

Supplier: Matrix Scientific

072495-250MG 072495-1G 072495-5G
102525-146EA 355.38 USD
102525-146 102525-148 102525-150
5-Bromo-3,7-dihydro-4H-pyrrolo[2,3-d]pyrimidin-4-one ≥95%
5-Bromo-3,7-dihydro-4H-pyrrolo[2,3-d]pyrimidin-4-one

Hazard: Irritant

Formula: C₆H₄BrN₃O
MDL Number: MFCD09999236
CAS Number: 22276-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR