6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one ≥95%

Supplier: Matrix Scientific

072494-1G 072494-5G
102525-142EA 176.92 USD
102525-142 102525-144
6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one ≥95%
6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one

Hazard: Irritant

Formula: C₈H₇BrN₂O
MDL Number: MFCD05664027
CAS Number: 129686-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR