Ethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride

072493-250MG 072493-1G 072493-5G
102525-134EA 569.23 USD
102525-134 102525-136 102525-138
Ethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride ≥95%
Ethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClN₃O₂
MW: 231.6794 g/mol
MDL Number: MFCD09259961
CAS Number: 623906-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR