6-Morpholinopyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Morpholinopyrimidin-4-amine, 4-Amino-6-morpholinopyrimidine

072491-1G 072491-5G 072491-10G
102525-122EA 569.23 USD
102525-122 102525-124 102525-126
6-Morpholinopyrimidin-4-amine ≥95%
6-Morpholinopyrimidin-4-amine

Hazard: Irritant

Formula: C₈H₁₂N₄O
MW: 180.2071 g/mol
MDL Number: MFCD09999228
CAS Number: 96225-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR