5-Bromo-6-chloro-4(3H)-pyrimidinone ≥95%

Supplier: Matrix Scientific

072488-250MG 072488-1G
102525-106EA 640 USD
102525-106 102525-108
5-Bromo-6-chloro-4(3H)-pyrimidinone ≥95%
5-Bromo-6-chloro-4(3H)-pyrimidinone

Hazard: Irritant

Formula: C₄H₂BrClN₂O
MW: 209.43 g/mol
MDL Number: MFCD09999225
CAS Number: 89089-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR