4-Amino-5-chloropyrimidine ≥95%

Supplier: Matrix Scientific

072485-250MG 072485-1G 072485-5G
102525-090EA 350.81 USD
102525-090 102525-092 102525-094
4-Amino-5-chloropyrimidine ≥95%
4-Amino-5-chloropyrimidine

Hazard: Irritant

Formula: C₄H₄ClN₃
MW: 129.5500 g/mol
MDL Number: MFCD00233964
CAS Number: 101257-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR