4-Aminoisoxazole hydrochloride 95

Supplier: Matrix Scientific

072484-250MG 072484-1G
102525-086EA 538.46 USD
102525-086 102525-088
4-Aminoisoxazole hydrochloride 95
4-Aminoisoxazole hydrochloride

Hazard: Irritant

Formula: C₃H₅ClN₂O
MW: 120.5400 g/mol
MDL Number: MFCD09992881
CAS Number: 108511-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR