4-Iodoisoxazole ≥95%

Supplier: Matrix Scientific

072483-1G 072483-5G
102525-080EA 235.38 USD
102525-080 102525-082
4-Iodoisoxazole ≥95%
4-Iodoisoxazole

Hazard: Irritant

Formula: C₃H₂INO
MW: 194.9600 g/mol
MDL Number: MFCD09999222
CAS Number: 847490-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR