4-Aminoisoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: Isoxazol-4-amine

072479-250MG 072479-1G 072479-5G
102525-056EA 321.58 USD
102525-056 102525-058 102525-060
4-Aminoisoxazole ≥95%
4-Aminoisoxazole

Hazard: Irritant/Store in Freezer

Formula: C₃H₄N₂O
MW: 84.0767 g/mol
MDL Number: MFCD08234629
CAS Number: 108511-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR