tert-Butyl-4-(4-bromo-2-cyanophenyl)piperazine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-(4-bromo-2-cyanophenyl)piperazine-1-carboxylate
072475-250MG
072475-1G
072475-5G
102525-036EA
355.38
USD
102525-036
102525-038
102525-040
tert-Butyl-4-(4-bromo-2-cyanophenyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(4-bromo-2-cyanophenyl)piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₀BrN₃O₂ MW: 366.2500 g/mol |
MDL Number:
MFCD09999215 CAS Number: 791846-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...