5-(4-Bromo-2-fluorophenyl)-1,3,4-thiadiazol-2-amine ≥95%

Supplier: Matrix Scientific

072473-250MG 072473-1G 072473-5G
102525-026EA 427.69 USD
102525-026 102525-028 102525-030
5-(4-Bromo-2-fluorophenyl)-1,3,4-thiadiazol-2-amine ≥95%
5-(4-Bromo-2-fluorophenyl)-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₈H₅BrFN₃S
MW: 274.11 g/mol
MDL Number: MFCD09999212
CAS Number: 299937-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR