4-Bromo-2-methoxybenzonitrile ≥95%

Supplier: Matrix Scientific

072472-1G 072472-5G 072472-10G
102525-020EA 15.38 USD
102525-020 102525-022 102525-024
4-Bromo-2-methoxybenzonitrile ≥95%
4-Bromo-2-methoxybenzonitrile

Hazard: Irritant

Formula: C₈H₆BrNO
MW: 212.05 g/mol
MDL Number: MFCD06738651
CAS Number: 330793-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR