3-Aminobenzo[d]isoxazole-6-carbonitrile ≥95%

Supplier: Matrix Scientific

072471-250MG 072471-1G
102525-016EA 390.77 USD
102525-016 102525-018
3-Aminobenzo[d]isoxazole-6-carbonitrile ≥95%
3-Aminobenzo[d]isoxazole-6-carbonitrile

Hazard: Irritant

Formula: C8H5N3O
MW: 159.1500 g/mol
MDL Number: MFCD09907912
CAS Number: 229623-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR