7-Bromoquinazoline-2,4-diamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Diamino-7-bromoquinazoline, DHFR-IN-3

072470-250MG 072470-1G 072470-5G
102525-010EA 352.31 USD
102525-010 102525-012 102525-014
7-Bromoquinazoline-2,4-diamine ≥95%
7-Bromoquinazoline-2,4-diamine

Hazard: Irritant

Formula: C₈H₇BrN₄
MDL Number: MFCD08706214
CAS Number: 137553-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR