2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide ≥95%

Supplier: Matrix Scientific

072469-250MG 072469-1G 072469-5G
102525-004EA 498.46 USD
102525-004 102525-006 102525-008
2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide ≥95%
2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide

Hazard: Irritant

Formula: C₆H₈Br₂N₂O
CAS Number: 443956-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR