3-Amino-4-pyridazinecarboxylic acid ≥95%

Supplier: Matrix Scientific

072468-250MG 072468-1G 072468-5G
102524-998EA 569.23 USD
102524-998 102525-000 102525-002
3-Amino-4-pyridazinecarboxylic acid ≥95%
3-Amino-4-pyridazinecarboxylic acid

Hazard: Irritant

Formula: C5H5N3O2
MW: 139.1100 g/mol
MDL Number: MFCD01318258
CAS Number: 21141-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR