2-Amino-4-methylpyrimidine-5-carbonitrile ≥95%
Supplier: Matrix Scientific
072465-250MG
072465-1G
102524-982EA
258.46
USD
102524-982
102524-984
2-Amino-4-methylpyrimidine-5-carbonitrile ≥95%
2-Amino-4-methylpyrimidine-5-carbonitrile
Hazard: Irritant
Formula:
C₆H₆N₄ MW: 134.1400 g/mol |
MDL Number:
MFCD09833141 CAS Number: 17321-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...