7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one ≥95%

Supplier: Matrix Scientific

072461-250MG 072461-1G
102524-962EA 783.08 USD
102524-962 102524-964
7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one ≥95%
7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one

Hazard: Irritant

Formula: C₆H₄BrN₃O
MW: 214.0195 g/mol
MDL Number: MFCD09999206
CAS Number: 93587-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR