5-Amino-2-bromopyrimidine ≥95%

Supplier: Matrix Scientific

072460-250MG 072460-1G 072460-5G
102524-956EA 692.31 USD
102524-956 102524-958 102524-960
5-Amino-2-bromopyrimidine ≥95%
5-Amino-2-bromopyrimidine

Hazard: Irritant

Formula: C₄H₄BrN₃
MW: 173.9987 g/mol
MDL Number: MFCD07778608
CAS Number: 56621-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR