5-Amino-4-bromopyrimidine ≥95%

Supplier: Matrix Scientific

072459-250MG 072459-1G 072459-5G
102524-950EA 2196.92 USD
102524-950 102524-952 102524-954
5-Amino-4-bromopyrimidine ≥95%
5-Amino-4-bromopyrimidine

Hazard: Irritant/Refrigerate

Formula: C₄H₄BrN₃
MDL Number: MFCD09999203
CAS Number: 849353-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR