6-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde ≥95%
Supplier: Matrix Scientific
072455-250MG
072455-1G
072455-5G
102524-930EA
769.23
USD
102524-930
102524-932
102524-934
6-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde ≥95%
6-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde
Hazard: Irritant
Formula:
C₈H₅BrN₂O MW: 225.0400 g/mol |
MDL Number:
MFCD09864909 CAS Number: 30384-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...