2-Bromo-6-methylpyrazino[2,3-b]pyrazine ≥95%

Supplier: Matrix Scientific

072450-250MG 072450-1G
102524-908EA 684.62 USD
102524-908 102524-910
2-Bromo-6-methylpyrazino[2,3-b]pyrazine ≥95%
2-Bromo-6-methylpyrazino[2,3-b]pyrazine

Hazard: Irritant

Formula: C₇H₅BrN₄
MDL Number: MFCD08706339
CAS Number: 91225-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR