2-(4-Methylpiperazin-1-yl)pyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

072449-250MG 072449-1G 072449-5G
102524-902EA 498.46 USD
102524-902 102524-904 102524-906
2-(4-Methylpiperazin-1-yl)pyrimidin-4-amine ≥95%

Hazard: Irritant

Formula: C₉H₁₅N₅
MDL Number: MFCD09999199
CAS Number: 57005-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR