2-Chloro-6-fluoroquinoxaline ≥95%
Supplier: Matrix Scientific
072446-250MG
072446-1G
072446-5G
102524-886EA
498.46
USD
102524-886
102524-888
102524-890
2-Chloro-6-fluoroquinoxaline ≥95%
2-Chloro-6-fluoroquinoxaline
Hazard: Irritant
Formula:
C₈H₄ClFN₂ MW: 182.5800 g/mol |
MDL Number:
MFCD09999192 CAS Number: 55687-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...