6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo[1,3]oxazolo[4,5-b]pyridin-2(3H)-one, 6-bromooxazolo[4,5-b]pyridin-2(3H)-one

072444-1G 072444-5G 072444-10G
102524-874EA 340 USD
102524-874 102524-876 102524-878
6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one ≥95%
6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one

Hazard: Irritant

Formula: C₆H₃BrN₂O₂
MW: 215.006 g/mol
MDL Number: MFCD08689590
CAS Number: 21594-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR