2-(Boc-amino)-4-bromopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (4-bromopyridin-2-yl)carbamate

072443-1G 072443-5G
102524-870EA 476.92 USD
102524-870 102524-872
2-(Boc-amino)-4-bromopyridine ≥95%
2-(Boc-amino)-4-bromopyridine

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O₂
MW: 273.13 g/mol
MDL Number: MFCD09037866
CAS Number: 207799-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR