2-Amino-4-bromothiazole ≥95%

Supplier: Matrix Scientific

072442-1G 072442-5G
102524-864EA 276.92 USD
102524-864 102524-866
2-Amino-4-bromothiazole ≥95%
2-Amino-4-bromothiazole

Hazard: Irritant

Formula: C₃H₃BrN₂S
MDL Number: MFCD06804547
CAS Number: 502145-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR