6-Bromo-2-chloroquinazoline ≥95%

Supplier: Matrix Scientific

072441-1G 072441-5G
102524-860EA 150.77 USD
102524-860 102524-862
6-Bromo-2-chloroquinazoline ≥95%
6-Bromo-2-chloroquinazoline

Hazard: Irritant

Formula: C₈H₄BrClN₂
MW: 243.4900 g/mol
MDL Number: MFCD09261000
CAS Number: 882672-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR