1-Benzyl-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

072439-250MG 072439-1G 072439-5G
102524-846EA 355.38 USD
102524-846 102524-848 102524-850
1-Benzyl-1H-pyrazole-4-carbonitrile ≥95%
1-Benzyl-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₁H₉N₃
CAS Number: 121358-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR