4-Chloropyrido[3,4-d]pyrimidine ≥95%

Supplier: Matrix Scientific

072438-250MG 072438-1G
102524-842EA 684.62 USD
102524-842 102524-844
4-Chloropyrido[3,4-d]pyrimidine ≥95%
4-Chloropyrido[3,4-d]pyrimidine

Hazard: Irritant

Formula: C₇H₄ClN₃
MW: 165.5798 g/mol
MDL Number: MFCD09999184
CAS Number: 51752-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR