6-Fluoro-1-naphthoic acid ≥95%

Supplier: Matrix Scientific

072434-250MG 072434-1G 072434-5G
102524-820EA 640 USD
102524-820 102524-822 102524-824
6-Fluoro-1-naphthoic acid ≥95%
6-Fluoro-1-naphthoic acid

Hazard: Irritant

Formula: C₁₁H₇FO₂
CAS Number: 575-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR