2-Bromo-5-phenyl-1,3,4-oxadiazole ≥95%

Supplier: Matrix Scientific

072432-250MG 072432-1G 072432-5G
102524-808EA 427.69 USD
102524-808 102524-810 102524-812
2-Bromo-5-phenyl-1,3,4-oxadiazole ≥95%
2-Bromo-5-phenyl-1,3,4-oxadiazole

Hazard: Irritant

Formula: C₈H₅BrN₂O
MW: 225.04 g/mol
MDL Number: MFCD06660932
CAS Number: 51039-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR