2-Hydroxypyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: Pyrazin-2(1H)-one, 2(1H)-Pyrazinone, 2-Pyrazinol

072430-1G 072430-5G
102524-798EA 15.38 USD
102524-798 102524-800
2-Hydroxypyrazine ≥95%
2-Hydroxypyrazine

Hazard: Irritant

Formula: C₄H₄N₂O
MW: 96.09 g/mol
MDL Number: MFCD00233966
CAS Number: 6270-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR