5-Ethylpiperidine-2,4-dione ≥95%

Supplier: Matrix Scientific

072428-250MG 072428-1G
102524-790EA 684.62 USD
102524-790 102524-792
5-Ethylpiperidine-2,4-dione ≥95%
5-Ethylpiperidine-2,4-dione

Hazard: Irritant

Formula: C7H11NO2
MW: 141.1700 g/mol
MDL Number: MFCD09999166
CAS Number: 73290-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR