Ethyl-2-chlorothiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

072426-1G 072426-5G
102524-780EA 29.23 USD
102524-780 102524-782
Ethyl-2-chlorothiazole-5-carboxylate ≥95%
Ethyl-2-chlorothiazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₆ClNO₂S
MDL Number: MFCD08703267
CAS Number: 81449-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR