2-Nitropyridine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

072425-250MG 072425-1G 072425-5G
102524-774EA 498.46 USD
102524-774 102524-776 102524-778
2-Nitropyridine-5-carboxylic acid ≥95%
2-Nitropyridine-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₄N₂O₄
MW: 168.1070 g/mol
MDL Number: MFCD04114183
CAS Number: 33225-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR