1'-Boc-1,2-dihydro-2-oxospiro[4H-3,1-benzoxazine-4,4'-piperidine] ≥95%

Supplier: Matrix Scientific

Synonyms: 1'-Boc-1,2-dihydro-2-oxo-spiro[4H-3,1-benzoxazine-4,4'-piperidine]

072421-250MG 072421-1G
102524-754EA 684.62 USD
102524-754 102524-756
1'-Boc-1,2-dihydro-2-oxospiro[4H-3,1-benzoxazine-4,4'-piperidine] ≥95%
1'-Boc-1,2-dihydro-2-oxospiro[4H-3,1-benzoxazine-4,4'-piperidine]

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MW: 318.3676 g/mol
MDL Number: MFCD10566068
CAS Number: 84060-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR