Spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one ≥95%

Supplier: Matrix Scientific

072420-250MG 072420-1G
102524-750EA 967.69 USD
102524-750 102524-752
Spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one ≥95%
Spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂
CAS Number: 84060-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR