6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one ≥95%

Supplier: Matrix Scientific

072418-250MG 072418-1G
102524-742EA 415.38 USD
102524-742 102524-744
6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one ≥95%
6-Fluorospiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-one

Hazard: Irritant

Formula: C₁₂H₁₃FN₂O₂
MW: 236.2400 g/mol
MDL Number: MFCD10566063
CAS Number: 92926-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR