5-Hydroxy-2H-benzo[b][1,4]oxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

072412-250MG 072412-1G
102524-712EA 498.46 USD
102524-712 102524-714
5-Hydroxy-2H-benzo[b][1,4]oxazin-3(4H)-one ≥95%
5-Hydroxy-2H-benzo[b][1,4]oxazin-3(4H)-one

Hazard: Irritant

Formula: C8H7NO3
CAS Number: 177210-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR