4-(Piperidin-4-ylmethyl)benzonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

072411-250MG 072411-1G 072411-5G
102524-706EA 498.46 USD
102524-706 102524-708 102524-710
4-(Piperidin-4-ylmethyl)benzonitrile hydrochloride ≥95%
4-(Piperidin-4-ylmethyl)benzonitrile hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂
CAS Number: 333987-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR