4-(2-Pyridinyl)-4-piperidinol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-4-pyrid-2-yl(piperidine)

072410-250MG 072410-1G 072410-5G
102524-700EA 423.69 USD
102524-700 102524-702 102524-704
4-(2-Pyridinyl)-4-piperidinol ≥95%
4-(2-Pyridinyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₀H₁₄N₂O
CAS Number: 50461-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR