3-(4-Piperidinyl)-1,3-benzoxazol-2(3H)-one ≥95%

Supplier: Matrix Scientific

072409-250MG 072409-1G 072409-5G
102524-694EA 498.46 USD
102524-694 102524-696 102524-698
3-(4-Piperidinyl)-1,3-benzoxazol-2(3H)-one ≥95%
3-(4-Piperidinyl)-1,3-benzoxazol-2(3H)-one

Hazard: Irritant

Formula: C12H14N2O2
CAS Number: 215878-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR