tert-Butyl-4-guanidinopiperidine-1-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

072407-250MG 072407-1G
102524-686EA 924.62 USD
102524-686 102524-688
tert-Butyl-4-guanidinopiperidine-1-carboxylate hydrochloride ≥95%
tert-Butyl-4-guanidinopiperidine-1-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₄O₂
CAS Number: 885049-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR