Pyrazine-2-carboximidamide hydrochloride ≥95%
Supplier: Matrix Scientific
072406-1G
072406-5G
072406-10G
102524-680EA
533.85
USD
102524-680
102524-682
102524-684
Pyrazine-2-carboximidamide hydrochloride ≥95%
Pyrazine-2-carboximidamide hydrochloride
Hazard: Irritant
Formula:
C5H7ClN4 MW: 158.5900 g/mol |
MDL Number:
MFCD05663360 CAS Number: 138588-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...